3-Bromo-5-fluorophenyl acetate ≥97% research grade

Supplier: AOBChem USA

Warning

107963-1G 107963-25G 107963-5G
76906-496EA 82.76 USD
76906-496 76906-498 76906-500
3-Bromo-5-fluorophenyl acetate ≥97% research grade
3-Bromo-5-fluorophenyl acetate
Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
CAS Number: 1260846-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR