(3-Formyl-4-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Formyl-4-methylphenylboronic acid

10553-25G 10553-5G
76906-446EA 190.97 USD
76906-446 76906-448
(3-Formyl-4-methylphenyl)boronic acid ≥97% research grade
(3-Formyl-4-methylphenyl)boronic acid
Formula: C₈H₉BO₃
MW: 163.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08274476
CAS Number: 1106869-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR