4-[Methoxy(methyl)carbamoyl]benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(N,O-Dimethylhydroxylaminocarbonyl)phenylboronic acid, 4-Borono-(N-methoxy-N-methyl)benzamide

10546-25G 10546-5G
76906-440EA 204.76 USD
76906-440 76906-442
4-[Methoxy(methyl)carbamoyl]benzeneboronic acid ≥97%
4-[Methoxy(methyl)carbamoyl]benzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BNO₄
Storage Temperature: Ambient
MDL Number: MFCD04115671
CAS Number: 179055-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR