4-(3-Chlorophenoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

104778-1G 104778-25G 104778-500MG 104778-5G
76906-408EA 135.8 USD
76906-408 76906-410 76906-412 76906-414
4-(3-Chlorophenoxy)benzaldehyde ≥97%
4-(3-Chlorophenoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07365211
CAS Number: 164522-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR