4-(Benzyloxy)-5-bromo-N,N-dimethylpyrimidin-2-amine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Benzyloxy-5-bromo-2-(N,N-dimethylamino)pyrimidine

10436-1G 10436-25G 10436-5G
76906-390EA 73.21 USD
76906-390 76906-392 76906-394
4-(Benzyloxy)-5-bromo-N,N-dimethylpyrimidin-2-amine ≥97%
4-(Benzyloxy)-5-bromo-N,N-dimethylpyrimidin-2-amine
Formula: C₁₃H₁₄BrN₃O
MW: 308.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458469
CAS Number: 205672-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR