4-Bromo-2-chlorophenetole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-chloro-ethoxybenzene, 4-Bromo-2-chloro-1-ethoxybenzene

10369-25G 10369-5G
76906-376EA 230.22 USD
76906-376 76906-378
4-Bromo-2-chlorophenetole ≥97%
4-Bromo-2-chlorophenetole

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835099
CAS Number: 279261-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR