4-Benzyloxy-2-formylphenylboronic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(4-(Benzyloxy)-2-formylphenyl)boronic acid
10308-25G
10308-5G
76906-356EA
279.02
USD
76906-356
76906-358
4-Benzyloxy-2-formylphenylboronic acid ≥97% research grade
4-Benzyloxy-2-formylphenylboronic acid
Formula:
C₁₄H₁₃BO₄ Storage Temperature: Ambient |
MDL Number:
MFCD02179486 CAS Number: 139962-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...