3-Methoxy-4-(methoxycarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-carboxy-3-methoxyphenyl boronic acid methyl ester, 3-Methoxy-4-methoxycarbonylphenylboronic acid, Benzoic acid, 4-borono-2-methoxy-, methyl ester, 3-Methoxy-4-(methoxycarbonyl)benzeneboronic acid

10280-25G 10280-5G
76906-340EA 204.76 USD
76906-340 76906-342
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid ≥97% research grade
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₅
MW: 209.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366483
CAS Number: 603122-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR