3-(Piperidine-1-carbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10255-25G 10255-5G
76906-322EA 310.85 USD
76906-322 76906-324
3-(Piperidine-1-carbonyl)phenylboronic acid ≥97%
3-(Piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₆BNO₃
MW: 233.0713 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115686
CAS Number: 850568-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR