3-Bromo-5-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

10140-25G 10140-5G
76906-274EA 410.57 USD
76906-274 76906-276
3-Bromo-5-ethoxypyridine ≥97%
3-Bromo-5-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234116
CAS Number: 17117-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR