1-Chloro-2,4-difluoro-5-methoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

97680-1G 97680-25G 97680-500MG 97680-5G
76893-988EA 424.36 USD
76893-988 76893-990 76893-992 76893-994
1-Chloro-2,4-difluoro-5-methoxybenzene ≥97% research grade
1-Chloro-2,4-difluoro-5-methoxybenzene
Formula: C₇H₅ClF₂O
MW: 178.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18415547
CAS Number: 1261628-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR