(4-cyano-2-(trifluoromethyl)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (4-Cyano-2-(trifluoromethyl)phenyl)boronic acid

95923-1G 95923-500MG 95923-5G 95923-25G
76893-978EA 554.86 USD
76893-978 76893-980 76893-982 77657-106
(4-cyano-2-(trifluoromethyl)phenyl)boronic acid ≥97% research grade
(4-cyano-2-(trifluoromethyl)phenyl)boronic acid
Formula: C₈H₅BF₃NO₂
MW: 214.937 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28782839
CAS Number: 2407353-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR