1-Fluoro-4-iodo-2-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

94574-1G 94574-25G 94574-5G
76893-964EA 75.33 USD
76893-964 76893-966 76893-968
1-Fluoro-4-iodo-2-isopropylbenzene ≥97%
1-Fluoro-4-iodo-2-isopropylbenzene
Formula: C₉H₁₀FI
MW: 264.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29091196
CAS Number: 1369775-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR