1-(2-Fluoro-4-methylphenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

93392-1G 93392-25G 93392-500MG 93392-5G
76893-948EA 321.46 USD
76893-948 76893-950 76893-952 76893-954
1-(2-Fluoro-4-methylphenyl)cyclobutanol ≥97%
1-(2-Fluoro-4-methylphenyl)cyclobutanol
Formula: C₁₁H₁₃FO
MW: 180.219 g/mol
Storage Temperature: Refrigerator
CAS Number: 2167603-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR