(5-Bromo-8-fluoroquinolin-7-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

92676-1G 92676-25G 92676-500MG 92676-5G
76893-936EA 380.87 USD
76893-936 76893-938 76893-940 76893-942
(5-Bromo-8-fluoroquinolin-7-yl)boronic acid ≥95%
(5-Bromo-8-fluoroquinolin-7-yl)boronic acid
Formula: C₉H₆BBrFNO₂
MW: 269.863 g/mol
Storage Temperature: Ambient
CAS Number: 2377606-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR