2-(3-Chlorophenyl)-2-oxoacetic acid ≥97%

Supplier: AOBChem USA

Warning

92491-1G 92491-25G 92491-500MG 92491-5G
76893-920EA 217.49 USD
76893-920 76893-922 76893-924 76893-926
2-(3-Chlorophenyl)-2-oxoacetic acid ≥97%
2-(3-Chlorophenyl)-2-oxoacetic acid
Formula: C₈H₅ClO₃
Storage Temperature: Ambient
CAS Number: 26767-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR