2,2,2-trifluoro-N-(4-isopropylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

91362-1G 91362-25G 91362-500MG 91362-5G
76893-896EA 213.25 USD
76893-896 76893-898 76893-900 76893-902
2,2,2-trifluoro-N-(4-isopropylphenyl)acetamide ≥97%
2,2,2-trifluoro-N-(4-isopropylphenyl)acetamide
Formula: C₁₁H₁₂F₃NO
Storage Temperature: Ambient
CAS Number: 75999-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR