1-(2,2-Dibromovinyl)-3,5-Difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

90037-1G 90037-25G 90037-500MG 90037-5G
76893-868EA 393.6 USD
76893-868 76893-870 76893-872 76893-874
1-(2,2-Dibromovinyl)-3,5-Difluorobenzene ≥95%
1-(2,2-Dibromovinyl)-3,5-Difluorobenzene
Formula: C₈H₄Br₂F₂
MW: 297.922 g/mol
Storage Temperature: Refrigerator
CAS Number: 1879225-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR