1-Bromo-4-(2,2-dibromovinyl)-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

89706-1G 89706-25G 89706-500MG 89706-5G
76893-842EA 381.93 USD
76893-842 76893-844 76893-846 76893-848
1-Bromo-4-(2,2-dibromovinyl)-2-fluorobenzene ≥95%
1-Bromo-4-(2,2-dibromovinyl)-2-fluorobenzene
Formula: C₈H₄Br₃F
MW: 358.828 g/mol
Storage Temperature: Refrigerator
CAS Number: 1865598-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR