1-(3-(1,3-dioxolan-2-yl)phenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89168-25G 89168-5G 89168-1G 89168-500MG
76893-830EA 616.39 USD
76893-830 76893-832 77640-148 77652-154
1-(3-(1,3-dioxolan-2-yl)phenyl)cyclohexanol ≥97%
1-(3-(1,3-dioxolan-2-yl)phenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₃
Storage Temperature: Refrigerator
MDL Number: MFCD34817905
CAS Number: 224031-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR