(6-Methylbenzo[d][1,3]dioxol-5-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

87617-1G 87617-500MG 87617-5G
76893-794EA 827.51 USD
76893-794 76893-796 76893-798
(6-Methylbenzo[d][1,3]dioxol-5-yl)boronic acid ≥97%
(6-Methylbenzo[d][1,3]dioxol-5-yl)boronic acid
Formula: C₈H₉BO₄
MW: 179.966 g/mol
Storage Temperature: Ambient
CAS Number: 148839-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR