1-(5-Chloro-2-fluorophenyl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

85997-1G 85997-25G 85997-500MG 85997-5G
76893-764EA 334.19 USD
76893-764 76893-766 76893-768 76893-770
1-(5-Chloro-2-fluorophenyl)cyclopentanol ≥97%
1-(5-Chloro-2-fluorophenyl)cyclopentanol
Formula: C₁₁H₁₂ClFO
MW: 214.664 g/mol
Storage Temperature: Ambient
CAS Number: 1516440-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR