1-(2,4-Difluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85820-1G 85820-500MG 85820-5G
76893-750EA 560.16 USD
76893-750 76893-752 76893-754
1-(2,4-Difluorophenyl)cyclobutanol ≥95%
1-(2,4-Difluorophenyl)cyclobutanol
Formula: C₁₀H₁₀F₂O
MW: 184.183 g/mol
Storage Temperature: Refrigerator
CAS Number: 1505928-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR