1-Bromo-3-(1,1-difluoroethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

84010-1G 84010-25G 84010-5G
76893-716EA 62.6 USD
76893-716 76893-718 76893-720
1-Bromo-3-(1,1-difluoroethyl)benzene ≥97%
1-Bromo-3-(1,1-difluoroethyl)benzene
Formula: C₈H₇BrF₂
Storage Temperature: Refrigerator
MDL Number: MFCD11007736
CAS Number: 445303-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR