1-Ethoxy-2-ethylbenzene ≥97%

Supplier: AOBChem USA

Warning

83743-25G 83743-5G
76893-706EA 460.44 USD
76893-706 76893-708
1-Ethoxy-2-ethylbenzene ≥97%
1-Ethoxy-2-ethylbenzene
Formula: C₁₀H₁₄O
MW: 150.218 g/mol
Storage Temperature: Ambient
CAS Number: 29643-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR