1,3-Dibromo-2-(2-bromoethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

82363-1G 82363-25G 82363-500MG 82363-5G
76893-682EA 151.71 USD
76893-682 76893-684 76893-686 76893-688
1,3-Dibromo-2-(2-bromoethoxy)benzene ≥95%
1,3-Dibromo-2-(2-bromoethoxy)benzene
Formula: C₈H₇Br₃O
Storage Temperature: Refrigerator
CAS Number: 206347-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR