(2,6-Dibromo-4-(Trifluoromethyl)Phenyl)(Methyl)Sulfane ≥95%

Supplier: AOBChem USA

Warning

80361-25G 80361-5G
76893-642EA 204.76 USD
76893-642 76893-644
(2,6-Dibromo-4-(Trifluoromethyl)Phenyl)(Methyl)Sulfane ≥95%
(2,6-Dibromo-4-(Trifluoromethyl)Phenyl)(Methyl)Sulfane
Formula: C₈H₅Br₂F₃S
Storage Temperature: Ambient
CAS Number: 2710880-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR