1-Bromo-4-(4-nitrophenoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

80054-1G 80054-25G 80054-500MG 80054-5G
76893-626EA 170.81 USD
76893-626 76893-628 76893-630 76893-632
1-Bromo-4-(4-nitrophenoxy)benzene ≥97%
1-Bromo-4-(4-nitrophenoxy)benzene
Formula: C₁₂H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD00222614
CAS Number: 21969-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR