1-(4-Bromobenzoyl)pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromophenyl)(pyrrolidin-1-yl)methanone

79963-25G 79963-5G
76893-616EA 222.79 USD
76893-616 76893-618
1-(4-Bromobenzoyl)pyrrolidine ≥97%
1-(4-Bromobenzoyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
MW: 254.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00595283
CAS Number: 5543-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR