1-(2-Isopropoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
78857-25G
78857-5G
76893-594EA
488.02
USD
76893-594
76893-596
1-(2-Isopropoxyphenyl)ethanone ≥97%
1-(2-Isopropoxyphenyl)ethanone
Formula:
C₁₁H₁₄O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01923827 CAS Number: 70201-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...