1,4-Dimethoxy-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

78643-1G 78643-25G 78643-500MG 78643-5G
76893-568EA 433.91 USD
76893-568 76893-570 76893-572 76893-574
1,4-Dimethoxy-2-(trifluoromethyl)benzene ≥97%
1,4-Dimethoxy-2-(trifluoromethyl)benzene
Formula: C₉H₉F₃O₂
MW: 206.162 g/mol
Storage Temperature: Ambient
CAS Number: 84355-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR