2,4-Dichloro-3-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

78561-1G 78561-25G 78561-500MG 78561-5G
76893-556EA 173.99 USD
76893-556 76893-558 76893-560 76893-562
2,4-Dichloro-3-ethoxybenzaldehyde ≥97%
2,4-Dichloro-3-ethoxybenzaldehyde
Formula: C₉H₈Cl₂O₂
MW: 219.065 g/mol
Storage Temperature: Refrigerator
CAS Number: 2385723-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR