1-Chloro-2-fluoro-3-iodo-4-methoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
77881-1G
77881-25G
77881-500MG
77881-5G
76893-526EA
177.18
USD
76893-526
76893-528
76893-530
76893-532
1-Chloro-2-fluoro-3-iodo-4-methoxybenzene ≥97%
1-Chloro-2-fluoro-3-iodo-4-methoxybenzene
Formula:
C₇H₅ClFIO MW: 286.47 g/mol Storage Temperature: Ambient |
CAS Number:
1402004-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...