1-(2,6-Difluorophenyl)ethanol ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-alpha-methylbenzyl alcohol

76198-25G 76198-5G
76893-490EA 209 USD
76893-490 76893-492
1-(2,6-Difluorophenyl)ethanol ≥95% research grade
1-(2,6-Difluorophenyl)ethanol
Formula: C₈H₈F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00274321
CAS Number: 87327-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR