2-(Methylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

75999-1G 75999-25G 75999-5G
76893-464EA 99.73 USD
76893-464 76893-466 76893-468
2-(Methylthio)benzonitrile ≥97%
2-(Methylthio)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₇NS
Storage Temperature: Ambient
MDL Number: MFCD00015557
CAS Number: 6609-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR