5'-Chloro-2'-hydroxy-4'-methoxyacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(5-Chloro-2-hydroxy-4-methoxyphenyl)ethanone

75703-1G 75703-25G 75703-500MG 75703-5G
76893-450EA 260.99 USD
76893-450 76893-452 76893-454 76893-456
5'-Chloro-2'-hydroxy-4'-methoxyacetophenone ≥97%
5'-Chloro-2'-hydroxy-4'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD03412058
CAS Number: 116265-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR