1-Bromo-2-fluoro-4-iodo-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

75348-1G 75348-25G 75348-500MG 75348-5G
76893-434EA 217.49 USD
76893-434 76893-436 76893-438 76893-440
1-Bromo-2-fluoro-4-iodo-5-methoxybenzene ≥97%
1-Bromo-2-fluoro-4-iodo-5-methoxybenzene
Formula: C₇H₅BrFIO
MW: 330.9208 g/mol
Storage Temperature: Ambient
CAS Number: 1447671-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR