N-(2-Fluorobenzoyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluorophenyl)(morpholino)methanone

75080-25G 75080-5G
76893-422EA 307.67 USD
76893-422 76893-424
N-(2-Fluorobenzoyl)morpholine ≥97%
N-(2-Fluorobenzoyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₂
MW: 209.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00120209
CAS Number: 1978-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR