1-Bromo-2,3-dichloro-4-(2-methylpropoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

74728-1G 74728-25G 74728-500MG 74728-5G
76893-406EA 168.69 USD
76893-406 76893-408 76893-410 76893-412
1-Bromo-2,3-dichloro-4-(2-methylpropoxy)benzene ≥95%
1-Bromo-2,3-dichloro-4-(2-methylpropoxy)benzene
Formula: C₁₀H₁₁BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD29043927
CAS Number: 1881329-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR