1-Bromo-5-fluoro-2,3-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

74152-1G 74152-25G 74152-500MG 74152-5G
76893-392EA 397.84 USD
76893-392 76893-394 76893-396 76893-398
1-Bromo-5-fluoro-2,3-dimethoxybenzene ≥95%
1-Bromo-5-fluoro-2,3-dimethoxybenzene
Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Refrigerator
CAS Number: 1784679-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR