(6-Bromo-2-chloro-3-methylphenoxy)(tert-butyl)dimethylsilane ≥95%

Supplier: AOBChem USA

Warning

73226-1G 73226-25G 73226-500MG 73226-5G
76893-350EA 155.96 USD
76893-350 76893-352 76893-354 76893-356
(6-Bromo-2-chloro-3-methylphenoxy)(tert-butyl)dimethylsilane ≥95%
(6-Bromo-2-chloro-3-methylphenoxy)(tert-butyl)dimethylsilane
Formula: C₁₃H₂₀BrClOSi
MW: 335.74 g/mol
Storage Temperature: Ambient
CAS Number: 2807471-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR