1-tert-Butyl-1H-pyrazole-4-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-(tert-Butyl)-1H-pyrazole-4-carboxylic acid
72834-1G
72834-25G
72834-500MG
72834-5G
76893-338EA
155.96
USD
76893-338
76893-340
76893-342
76893-344
1-tert-Butyl-1H-pyrazole-4-carboxylic acid ≥97%
1-tert-Butyl-1H-pyrazole-4-carboxylic acid
Formula:
C₈H₁₂N₂O₂ MW: 168.19 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09934934 CAS Number: 950858-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...