(4'-(Hydroxymethyl)-[1,1'-biphenyl]-4-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

72515-1G 72515-25G 72515-500MG 72515-5G
76893-296EA 434.97 USD
76893-296 76893-298 76893-300 76893-302
(4'-(Hydroxymethyl)-[1,1'-biphenyl]-4-yl)boronic acid ≥95%
(4'-(Hydroxymethyl)-[1,1'-biphenyl]-4-yl)boronic acid
Formula: C₁₃H₁₃BO₃
MW: 228.0515 g/mol
Storage Temperature: Ambient
CAS Number: 1315259-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR