1-(3,4-Dimethoxyphenyl)-2,2-difluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
71616-1G
71616-25G
71616-500MG
71616-5G
76893-282EA
424.36
USD
76893-282
76893-284
76893-286
76893-288
1-(3,4-Dimethoxyphenyl)-2,2-difluoroethanone ≥97%
1-(3,4-Dimethoxyphenyl)-2,2-difluoroethanone
Formula:
C₁₀H₁₀F₂O₃ MW: 216.181 g/mol Storage Temperature: Ambient |
CAS Number:
1271476-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...