1-(3-Hydroxythiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

71473-1G 71473-500MG 71473-5G
76893-268EA 664.13 USD
76893-268 76893-270 76893-272
1-(3-Hydroxythiophen-2-yl)ethanone ≥97%
1-(3-Hydroxythiophen-2-yl)ethanone
Formula: C₆H₆O₂S
Storage Temperature: Refrigerator
CAS Number: 5556-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR