1,2-Difluoro-3-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

71372-1G 71372-25G 71372-5G 71372-500MG
76893-254EA 77.45 USD
76893-254 76893-256 76893-258 77647-926
1,2-Difluoro-3-iodo-4-methoxybenzene ≥97%
1,2-Difluoro-3-iodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
MW: 270.0152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261400
CAS Number: 936498-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR