1-Methyl-4-(3-nitrophenyl)piperazine ≥97%

Supplier: AOBChem USA

Warning

69615-25G 69615-5G
76893-246EA 233.4 USD
76893-246 76893-248
1-Methyl-4-(3-nitrophenyl)piperazine ≥97%
1-Methyl-4-(3-nitrophenyl)piperazine
Formula: C₁₁H₁₅N₃O₂
MW: 221.256 g/mol
Storage Temperature: Ambient
CAS Number: 148546-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR