1-Bromo-2,4-difluoro-3-iodobenzene ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
69482-1G
69482-25G
69482-500MG
69482-5G
76893-230EA
147.47
USD
76893-230
76893-232
76893-234
76893-236
1-Bromo-2,4-difluoro-3-iodobenzene ≥97% research grade
1-Bromo-2,4-difluoro-3-iodobenzene
Formula:
C₆H₂BrF₂I MW: 318.88 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12922602 CAS Number: 1208075-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...