(5-Bromo-2,4-Difluorophenyl)(Methyl)Sulfane 95% research grade

Supplier: AOBChem USA

Warning

69314-1G 69314-500MG 69314-5G 69314-25G
76893-222EA 1490.57 USD
76893-222 76893-224 77630-614 77660-258
(5-Bromo-2,4-Difluorophenyl)(Methyl)Sulfane 95% research grade
(5-Bromo-2,4-Difluorophenyl)(Methyl)Sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrF₂S
Storage Temperature: Ambient
MDL Number: MFCD28131167
CAS Number: 1824059-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR