2-Bromo-4-chloro-5-fluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

68919-1G 68919-25G 68919-500MG 68919-5G
76893-190EA 245.07 USD
76893-190 76893-192 76893-194 76893-196
2-Bromo-4-chloro-5-fluoroiodobenzene ≥97%
2-Bromo-4-chloro-5-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
Storage Temperature: Ambient
MDL Number: MFCD12026232
CAS Number: 1067882-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR